Skip to Content
MilliporeSigma
All Photos(1)

Documents

W262235

Sigma-Aldrich

Lavender oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2622
MDL number:
UNSPSC Code:
12164502

biological source

Lavandula angustifolia

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 182.20

origin

eastern Europe origin

refractive index

n20/D 1.461 (lit.)

bp

204 °C (lit.)

density

0.879 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

lavender; herbaceous; floral

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatemeh Sheikhan et al.
Complementary therapies in clinical practice, 18(1), 66-70 (2011-12-27)
Post-episiotomy discomfort and its consequences can affect maternal quality of life and mental health as well as the mother and baby relationship. Complementary medicine is increasingly used and Lavender oil is frequently prescribed due to its antiseptic and healing properties.
D Tagreed Altaei
American journal of dentistry, 25(1), 39-43 (2012-05-09)
To determine the laboratory and clinical efficacy of lavender oil in the treatment of recurrent aphthous ulceration (RAU). This was a randomized double-blind, placebo-controlled study performed firstly to treat the induced ulcers by different methods in experimental animals (rabbits) treated
Dong Wang et al.
Molecules (Basel, Switzerland), 17(8), 9803-9817 (2012-08-17)
The air-dried aerial parts of Lavandula angustifolia Mill, a traditional Uygur herbal drug, is used as resuscitation-inducing therapy to treat neurodisfunctions, such as stroke. This study was designed to assess the neuroprotective effects of lavender oil against ischemia/reperfusion (IR) injury
Bengü Çetinkaya et al.
International journal of nursing practice, 18(2), 164-169 (2012-03-23)
The aim of this paper was to investigate the effect of aromatherapy massage using lavender oil as a possible treatment for this condition. This research was carried out on a group of 40 infants between 2 and 6 weeks of
P A R Hawken et al.
Physiology & behavior, 105(5), 1117-1123 (2011-12-24)
Growing concerns about the risk of addiction to benzodiazepines have led to increasing interest in alternative therapies to treat anxiety and depression. Lavender oil (Lavendula augustifolia) is reportedly anxiolytic in a number of species but little is known about how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service