Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W262000

Sigma-Aldrich

Lavender absolute

natural

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2620
MDL number:
UNSPSC Code:
12164502

biological source

Lavandula angustifolia L.

grade

natural

reg. compliance

FDA 21 CFR 182.20

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

herbaceous

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Tagreed Altaei
American journal of dentistry, 25(1), 39-43 (2012-05-09)
To determine the laboratory and clinical efficacy of lavender oil in the treatment of recurrent aphthous ulceration (RAU). This was a randomized double-blind, placebo-controlled study performed firstly to treat the induced ulcers by different methods in experimental animals (rabbits) treated
Serap Ejder Apay et al.
Pain management nursing : official journal of the American Society of Pain Management Nurses, 13(4), 236-240 (2012-11-20)
The purpose of this study was to investigate the effect of aromatherapy massage on dysmenorrhea. The study used a quasiexperimental design with the subjects as their own control. Every participant applied both aromatherapy massage with lavender oil and placebo massage
Mizuho Takahashi et al.
Natural product communications, 6(11), 1769-1774 (2012-01-10)
Essential oils have traditionally been used for decades to alleviate the symptoms of various mental problems. In terms of anxiolytic-like properties, lavender oil is probably the most commonly used and best-studied essential oil. Although there is compositional variance among the
Payam Sasannejad et al.
European neurology, 67(5), 288-291 (2012-04-21)
Lavender essential oil has been used as an anxiolytic drug, a mood stabilizer, a sedative, spasmolytic, antihypertensive, antimicrobial, analgesic agent as well as a wound healing accelerator. We have studied for the first time the efficacy of lavender essential oil
Buğra Ocak
Journal of environmental management, 100, 22-28 (2012-03-01)
In the world, approximately 600,000 metric tonnes of chromium-containing solid wastes are generated by the leather industry each year. Environmental concerns and escalating landfill costs are becoming increasingly serious problems to the leather industry and seeking solutions to these problems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service