Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W257615

Sigma-Aldrich

Hexyl propionate

natural (US), ≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2(CH2)5CH3
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2576
EC Number:
Council of Europe no.:
420
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.139
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural (US)

Quality Level

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

assay

≥97%

refractive index

n20/D 1.413 (lit.)

bp

73-74 °C/10 mmHg (lit.)

density

0.871 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

green; musty; fruity; pear

SMILES string

CCCCCCOC(=O)CC

InChI

1S/C9H18O2/c1-3-5-6-7-8-11-9(10)4-2/h3-8H2,1-2H3

InChI key

GOKKOFHHJFGZHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexyl propionate is a volatile flavor compound mainly found in different varieties of apple.[1] It also occurs in muskmelon.[2]

Other Notes

Natural occurrence: Apple, hops, apricot, melon, passion fruit, plum, gruyere cheese, chamomile oil.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composition of volatile compounds of apple fruits in relation to cultivars.
Kakiuchi N, et al.
Journal of the Japanese Society for Horticultural Science, 55(3), 280-289 (1986)
Volatile components of muskmelon fruit.
Kemp TR, et al.
Journal of Agricultural and Food Chemistry, 20(2), 196-198 (1972)
Rachel S Leisso et al.
Physiologia plantarum, 153(2), 204-220 (2014-06-20)
'Soggy breakdown' (SB) is an internal flesh disorder of 'Honeycrisp' apple (Malus × domestica Borkh.) fruit that occurs during low temperature storage. The disorder is a chilling injury (CI) in which visible symptoms typically appear after several weeks of storage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service