Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W256230

Sigma-Aldrich

trans-2-Hexen-1-ol

natural, 97%, FG

Synonym(s):

(E)-2-Hexen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH2OH
CAS Number:
Molecular Weight:
100.16
FEMA Number:
2562
Beilstein/REAXYS Number:
1719709
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.157
NACRES:
NA.21
organoleptic:
fresh; grassy; green; leafy
grade:
FG
Fragrance grade
Halal
Kosher
natural
agency:
follows IFRA guidelines
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

assay

97%

refractive index

n20/D 1.438 (lit.)

bp

158-160 °C (lit.)

density

0.849 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

fresh; grassy; green; leafy

SMILES string

[H]\C(CO)=C(\[H])CCC

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h4-5,7H,2-3,6H2,1H3/b5-4+

InChI key

ZCHHRLHTBGRGOT-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Natural occurrence: Raspberry, orange, apple, green tea, kiwi, peach, strawberry and tomato.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

129.2 °F - closed cup

flash_point_c

54 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Zhao et al.
Food research international (Ottawa, Ont.), 139, 109841-109841 (2021-01-30)
The commercial kiwifruit juice is deficient in a theoretical basis for the control of aroma characteristics during sterilization. To investigate the different sterilization methods on the aroma of kiwifruit juice, three sterilized kiwifruit juice samples, including pasteurization (PS), high temperature
Zuobing Xiao et al.
Journal of agricultural and food chemistry, 67(32), 8926-8937 (2019-07-30)
The volatile compounds found in mangoes (Mangifera indica L.) obtained from three cultivars (TN, Tainong no. 1; KT, Keitt; and ZL, Zill; grown in China) were comprehensively analyzed by gas chromatography-olfactometry (GC-O), gas chromatography-mass spectrometry (GC-MS), and GC-flame photometric detection
Xiaobing Pang
Journal of environmental sciences (China), 32, 196-206 (2015-06-05)
Green leaf volatiles (GLVs) emitted by plants after stress or damage induction are a major part of biogenic volatile organic compounds (BVOCs). Proton transfer reaction time-of-flight mass spectrometry (PTR-TOF-MS) is a high-resolution and sensitive technique for in situ GLV analyses
Béla P Molnár et al.
Frontiers in physiology, 9, 323-323 (2018-04-19)
Insects use sensitive olfactory systems to detect relevant host volatiles and avoid unsuitable hosts in a complex environmental odor landscape. Insects with short lifespans, such as gall midges (Diptera: Cecidomyiidae), are under strong selection pressure to detect and locate suitable
Kye Chung Park et al.
Journal of insect physiology, 78, 33-46 (2015-05-06)
Phytophagous insects detect volatile compounds produced by host and non-host plants, using species-specific sets of olfactory receptor neurons (ORNs). To investigate the relationship between the range of host plants and the profile of ORNs, single sensillum recordings were carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service