Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W253006

Sigma-Aldrich

Grapefruit oil, Florida

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2530
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Citrus paradisi Macf.

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D +92°, neat

origin

Florida origin

refractive index

n20/D 1.475

bp

171 °C

density

0.85 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

grapefruit

Preparation Note

Extraction method: cold pressed

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

131.0 °F - closed cup

flash_point_c

55 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service