Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W246900

Sigma-Aldrich

Eugenyl acetate

≥98%, FCC

Synonym(s):

4-Allyl-2-methoxyphenyl acetate, O-Acetyleugenol, Eugenol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
FEMA Number:
2469
EC Number:
Council of Europe no.:
210
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Flavis number:
9.020

biological source

synthetic

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

assay

≥98%

refractive index

n20/D 1.518 (lit.)

bp

281-286 °C (lit.)

density

1.079 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

COc1cc(CC=C)ccc1OC(C)=O

InChI

1S/C12H14O3/c1-4-5-10-6-7-11(15-9(2)13)12(8-10)14-3/h4,6-8H,1,5H2,2-3H3

InChI key

SCCDQYPEOIRVGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

287.6 °F - closed cup

flash_point_c

142 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service