Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W246611

Sigma-Aldrich

Eucalyptus oil

80-85%, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2466
EC Number:
UNSPSC Code:
12164502
NACRES:
NA.21
organoleptic:
herbaceous
grade:
FG
Kosher
food allergen:
no known allergens

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

assay

80-85%

origin

China origin

refractive index

n20/D 1.46

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

herbaceous

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3

InChI key

WEEGYLXZBRQIMU-UHFFFAOYSA-N

Preparation Note

Extraction method: steam distillation

pictograms

Flame

signalword

Warning

hcodes

pcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

134.8 °F - closed cup

flash_point_c

57.1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service