W246018
Ethyl tiglate
≥98%
Synonym(s):
Ethyl trans-2-methyl-2-butenoate
About This Item
biological source
synthetic
grade
Halal
reg. compliance
FDA 21 CFR 172.515
assay
≥98%
may contain
<0.10% alpha-tocopherol, synthetic as stabilizer
refractive index
n20/D 1.435 (lit.)
bp
154-156 °C (lit.)
density
0.923 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
organoleptic
caramel; fruity; raspberry
SMILES string
CCOC(=O)\C(C)=C\C
InChI
1S/C7H12O2/c1-4-6(3)7(8)9-5-2/h4H,5H2,1-3H3/b6-4+
InChI key
OAPHLAAOJMTMLY-GQCTYLIASA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service