W244511
Ethyl myristate
natural, 98%, FG
Synonym(s):
Ethyl tetradecanoate, Myristic acid ethyl ester
About This Item
Fragrance grade
Kosher
natural
Recommended Products
grade
FG
Fragrance grade
Kosher
natural
Quality Level
agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
assay
98%
refractive index
n20/D 1.436 (lit.)
n20/D 1.436
bp
178-180 °C/12 mmHg (lit.)
mp
11-12 °C (lit.)
density
0.86 g/mL at 25 °C (lit.)
0.863 g/mL at 25 °C
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
organoleptic
waxy; sweet
SMILES string
CCCCCCCCCCCCCC(=O)OCC
InChI
1S/C16H32O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16(17)18-4-2/h3-15H2,1-2H3
InChI key
MMKRHZKQPFCLLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service