Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W242004

Sigma-Aldrich

Ethyl p-anisate

≥97%, FCC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2C2H5
CAS Number:
Molecular Weight:
180.20
FEMA Number:
2420
EC Number:
Council of Europe no.:
249
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.714

biological source

synthetic

grade

Halal

reg. compliance

FCC
FDA 21 CFR 172.515

assay

≥97%

refractive index

n20/D 1.524 (lit.)

bp

263 °C (lit.)

mp

7-8 °C (lit.)

density

1.103 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

anise; fruity

SMILES string

CCOC(=O)c1ccc(OC)cc1

InChI

1S/C10H12O3/c1-3-13-10(11)8-4-6-9(12-2)7-5-8/h4-7H,3H2,1-2H3

InChI key

FHUODBDRWMIBQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service