Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W241318

Sigma-Aldrich

4-Ethoxybenzaldehyde

≥99%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CHO
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2413
Beilstein/REAXYS Number:
386863
EC Number:
Council of Europe no.:
626
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.056
NACRES:
NA.21
organoleptic:
spicy; sweet
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

assay

≥99%

refractive index

n20/D 1.559 (lit.)

bp

255 °C (lit.)

mp

13-14 °C (lit.)

density

1.08 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

spicy; sweet

SMILES string

[H]C(=O)c1ccc(OCC)cc1

InChI

1S/C9H10O2/c1-2-11-9-5-3-8(7-10)4-6-9/h3-7H,2H2,1H3

InChI key

JRHHJNMASOIRDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service