Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W239704

Sigma-Aldrich

1,3-Diphenyl-2-propanone

≥98%, FG

Synonym(s):

1,3-Diphenylacetone, Dibenzyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2CO
CAS Number:
Molecular Weight:
210.27
FEMA Number:
2397
Beilstein/REAXYS Number:
974767
EC Number:
Council of Europe no.:
2054
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.086
NACRES:
NA.21
organoleptic:
plastic; seedy
grade:
FG
Halal
Kosher
biological source:
synthetic
agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

assay

≥98%

bp

330 °C (lit.)

mp

32-34 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

plastic; seedy

SMILES string

O=C(Cc1ccccc1)Cc2ccccc2

InChI

1S/C15H14O/c16-15(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

YFKBXYGUSOXJGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Jin Lim et al.
Apoptosis : an international journal on programmed cell death, 20(8), 1087-1098 (2015-05-30)
Tunicamycin (TN), one of the endoplasmic reticulum stress inducers, has been reported to inhibit tumor cell growth and exhibit anticarcinogenic activity. However, the mechanism by which TN initiates apoptosis remains poorly understood. In the present study, we investigated the effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service