Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W238309

Sigma-Aldrich

Dillweed oil

natural, FCC

Synonym(s):

Anethum graveolens

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2383
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
organoleptic:
green; spicy; herbaceous
grade:
Kosher
natural
food allergen:
no known allergens

grade

Kosher
natural

Quality Level

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 184.1282

optical activity

[α]20/D +90°, neat

origin

Hungary origin

refractive index

n20/D 1.483 (lit.)

bp

189 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; spicy; herbaceous

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

127.9 °F - closed cup

flash_point_c

53.3 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service