Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W235407

Sigma-Aldrich

Cyclohexyl propionate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2C6H11
CAS Number:
Molecular Weight:
156.22
FEMA Number:
2354
EC Number:
Council of Europe no.:
421
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.140

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

assay

≥99%

refractive index

n20/D 1.441 (lit.)

bp

72-73 °C/10 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

banana; pineapple; sweet

SMILES string

CCC(=O)OC1CCCCC1

InChI

1S/C9H16O2/c1-2-9(10)11-8-6-4-3-5-7-8/h8H,2-7H2,1H3

InChI key

MAMMVUWCKMOLSG-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

154.4 °F - closed cup

flash_point_c

68 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service