Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W234605

Sigma-Aldrich

Cassis bourgens absolute

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2346
EC Number:
MDL number:
UNSPSC Code:
12164502

biological source

Ribes nigrum L.

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117
FDA 21 CFR 172.510

form

liquid

density

1.060 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; fruity; sulfurous

Biochem/physiol Actions

Taste at 5 ppm in 5% sugar solution.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

145.0 °F - closed cup

flash_point_c

62.78 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 24, 27-27 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service