Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W233609

Sigma-Aldrich

Costus oil

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2336
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
organoleptic:
woody; animal
grade:
FG
Kosher
natural
biological source:
Saussurea lappa Clarke
food allergen:
no known allergens

biological source

Saussurea lappa Clarke

Quality Level

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.510

origin

France origin

refractive index

n20/D 1.5180 (lit.)

density

1.002 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

woody; animal

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occupational erythema-multiforme-like dermatitis from sensitization to costus resinoid, followed by flare-up and systemic contact dermatitis from beta-cyclocostunolide in a chemistry student.
C J Le Coz et al.
Contact dermatitis, 44(5), 310-311 (2001-05-31)
I Takanami et al.
Gan no rinsho. Japan journal of cancer clinics, 29(11), 1319-1322 (1983-09-01)
Delayed hypersensitivity skin responses to costus root oil were measured in patients with colon, stomach, breast and lung cancer and in healthy controls. Cancer patients were staged according to tumor spread. Differences were demonstrated between cancer patients and controls. Patients
S Schauder
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 9(3), 182-187 (1998-09-24)
Although photosensitivity after photoallergy to topical phenothiazine antihistamines is well known, there have been no previous reports of dioxopromethazine inducing this phenomenon. A housewife used 0.5% dioxopromethazine in Prothanon gel for palpebral pruritus and developed severe dermatitis of the lower
C Benezra et al.
Contact dermatitis, 15(4), 223-230 (1986-10-01)
A molecular analysis of cross-reactions in costus-sensitive patients reveals that the most active sesquiterpene lactones are the ones with the lesser number of oxygenated substituents close to the alpha-methylene-gamma-butyrolactone ring. Cross-reactions with sesquiterpene lactones with different skeletons (germacranolide and guaianolide
I Takanami et al.
Gan to kagaku ryoho. Cancer & chemotherapy, 14(7), 2276-2279 (1987-07-01)
Costus root oil is a new primary sensitizer for examination of the delayed cell-mediated immune system. Histologic findings of skin producing a positive reaction to costus root oil show accumulation of lymphoid cells. If the delayed hypersensitivity reaction caused by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service