Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W232314

Sigma-Aldrich

Clove bud oil

Certified organic (NOP)

Synonym(s):

Clove oil, Eugenia spp., Oil of cloves

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2323
MDL number:
UNSPSC Code:
12164502
organoleptic:
clove; woody; spicy
grade:
Certified organic (NOP)
Halal
Kosher
food allergen:
no known allergens

grade

Certified organic (NOP)
Halal
Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 184.1257

origin

Madagascar origin

refractive index

n20/D 1.532 (lit.)

bp

251 °C (lit.)

density

1.04 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

clove; woody; spicy

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Essential oil used for its medicinal properties. Immunomodulator and anti-inflammatory.

Legal Information

Certified organic by MOSA

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

239.0 °F - closed cup

flash_point_c

115 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service