Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W230200

Sigma-Aldrich

trans-Cinnamyl isovalerate

≥95%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CO2CH2CH=CHC6H5
CAS Number:
Molecular Weight:
218.29
FEMA Number:
2302
Council of Europe no.:
454
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.133
NACRES:
NA.21
organoleptic:
spicy; fruity; woody; sour; wine-like
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

assay

≥95%

refractive index

n20/D 1.52 (lit.)

bp

313 °C (lit.)

density

0.992 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

spicy; fruity; woody; sour; wine-like

SMILES string

[H]\C(COC(=O)CC(C)C)=C(\[H])c1ccccc1

InChI

1S/C14H18O2/c1-12(2)11-14(15)16-10-6-9-13-7-4-3-5-8-13/h3-9,12H,10-11H2,1-2H3/b9-6+

InChI key

FOCMOGKCPPTERB-RMKNXTFCSA-N

Biochem/physiol Actions

Taste at 1-10 ppm

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 25, 72-72 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service