Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W229601

Sigma-Aldrich

trans-Cinnamyl butyrate

≥96%, FCC, FG

Synonym(s):

trans-Butyric acid cinnamyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH=CHC6H5
CAS Number:
Molecular Weight:
204.26
FEMA Number:
2296
Beilstein/REAXYS Number:
1956350
EC Number:
Council of Europe no.:
279
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.053
NACRES:
NA.21
organoleptic:
balsam; floral; fruity
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

assay

≥96%

refractive index

n20/D 1.526 (lit.)

bp

300 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

balsam; floral; fruity

SMILES string

CCCC(=O)OC\C=C\c1ccccc1

InChI

1S/C13H16O2/c1-2-7-13(14)15-11-6-10-12-8-4-3-5-9-12/h3-6,8-10H,2,7,11H2,1H3/b10-6+

InChI key

YZYPQKZWNXANRB-UXBLZVDNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service