Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W229411

Sigma-Aldrich

Cinnamyl alcohol

natural, 96%, FG

Synonym(s):

3-Phenyl-2-propen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCH2OH
CAS Number:
Molecular Weight:
134.18
FEMA Number:
2294
Beilstein/REAXYS Number:
1903999
EC Number:
Council of Europe no.:
65
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.017
NACRES:
NA.21
organoleptic:
cinnamon; green; balsamic; spicy
grade:
FG
Fragrance grade
Halal
Kosher
natural
agency:
follows IFRA guidelines
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

4.6 (vs air)

vapor pressure

<0.01 mmHg ( 25 °C)

assay

96%

bp

250 °C (lit.)

mp

30-33 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

cinnamyl alcohol

organoleptic

cinnamon; green; balsamic; spicy

SMILES string

OC\C=C\c1ccccc1

InChI

1S/C9H10O/c10-8-4-7-9-5-2-1-3-6-9/h1-7,10H,8H2/b7-4+

InChI key

OOCCDEMITAIZTP-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

258.8 °F - closed cup

flash_point_c

126 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Itai Chipinda et al.
Toxicology, 280(3), 135-143 (2010-12-18)
Consumer and medical products can contain leachable chemical allergens which can cause skin sensitization. Recent efforts have been directed at the development of non-animal based tests such as in vitro cell activation assays for the identification of skin sensitizers. Prohapten
Laura Fitzpatrick et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 40(1), 10-13 (2010-10-26)
Food-associated allergies, especially to benzoates and cinnamon-related compounds, have been associated with orofacial granulomatosis and both standard and urticarial patch testing have been used to detect such allergies. Elimination diets have also been shown to be effective in some patients.
Annette Zeller et al.
Chemical research in toxicology, 22(12), 1929-1937 (2009-11-17)
The metabolism of the potent carcinogen estragole was investigated in humans after consumption of fennel tea by analyses of its metabolites in blood plasma and urine. Stable isotope dilution assays based on LC-MS/MS detection revealed that 1'-hydroxylation of estragole happened
Carmen Martínez et al.
Journal of molecular graphics & modelling, 28(2), 196-201 (2009-08-04)
The free radical reactivity of monolignols can be explained in terms of the Fukui function and the local hard and soft acids and bases (HSAB) principle to determine the potential linkages among them for reactions involving free radicals. Our results
C S Letizia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 43(6), 837-866 (2005-04-07)
A toxicologic and dermatologic review of cinnamyl alcohol when used as a fragrance ingredient is presented.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service