Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W229208

Sigma-Aldrich

Cinnamon leaf oil

Certified organic (NOP)

Synonym(s):

Cinnamomum zeylanicum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2292
MDL number:
UNSPSC Code:
12164502

grade

Certified organic (NOP)
Halal
Kosher

reg. compliance

FDA 21 CFR 117

optical activity

[α]/D -2 to +2°, neat

refractive index

n20/D 1.5330
n20/D 1.535

bp

194-234 °C

density

1.041 g/mL at 25 °C
1.050 g/mL at 25 °C

application(s)

flavors and fragrances

organoleptic

spicy

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service