Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W229140

Sigma-Aldrich

Cinnamon bark oil

Certified organic (NOP)

Synonym(s):

Cassia oil, Cinnamomum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2291
MDL number:
UNSPSC Code:
12164502
organoleptic:
spicy; sweet
grade:
Certified organic (NOP)
Halal
Kosher
biological source:
Cinnamonum zeylanicum
food allergen:
no known allergens

biological source

Cinnamonum zeylanicum

grade

Certified organic (NOP)
Halal
Kosher

reg. compliance

FDA 21 CFR 117

refractive index

n20/D 1.545

density

0.970-1.100 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

spicy; sweet

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service