Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W223204

Sigma-Aldrich

Cananga oil

natural, FG

Synonym(s):

Cananga oil, Cananga Odorata

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2232
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

grade

FG
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D −24°, neat

composition

May contain estragol
May contain methyl eugenol

origin

Indonesia origin

refractive index

n20/D 1.5020 (lit.)

bp

264 °C (lit.)

density

0.911 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

woody; floral

Storage Class

10 - Combustible liquids not in Storage Class 3

wgk_germany

WGK 3

flash_point_f

190.0 °F - closed cup

flash_point_c

87.8 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service