Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W221600

Sigma-Aldrich

Butyl 10-undecenoate

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CH(CH2)8CO2(CH2)3CH3
CAS Number:
Molecular Weight:
240.38
FEMA Number:
2216
EC Number:
Council of Europe no.:
2103
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.238
NACRES:
NA.21
organoleptic:
butter; wine-like
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

assay

≥98%

refractive index

n20/D 1.4413 (lit.)

bp

249-254 °C/756 mmHg (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

butter; wine-like

SMILES string

CCCCOC(=O)CCCCCCCCC=C

InChI

1S/C15H28O2/c1-3-5-7-8-9-10-11-12-13-15(16)17-14-6-4-2/h3H,1,4-14H2,2H3

InChI key

GRAORJFMGCQWRN-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service