Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W219703

Sigma-Aldrich

Isobutyl formate

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2197
EC Number:
Council of Europe no.:
502
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.164
NACRES:
NA.21
organoleptic:
fruity; ethereal; sweet
grade:
FG
Halal
Kosher
biological source:
synthetic
agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

3.52 (vs air)

assay

≥95%

autoignition temp.

608 °F

expl. lim.

8.9 %

refractive index

n20/D 1.385 (lit.)

bp

98.4 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

fruity; ethereal; sweet

SMILES string

CC(C)COC=O

InChI

1S/C5H10O2/c1-5(2)3-7-4-6/h4-5H,3H2,1-2H3

InChI key

AVMSWPWPYJVYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 10 ppm

Other Notes

Natural occurrence: Apple, beer, grape brandy, pineapple, rum and vinegar.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

50.0 °F - closed cup

flash_point_c

10 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 105-105 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service