Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W212000

Sigma-Aldrich

Basil extract, linalol type

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2120
MDL number:
UNSPSC Code:
12164502
grade:
FG
Kosher
natural
biological source:
Ocimum basilicum L.
food allergen:
no known allergens

biological source

Ocimum basilicum L.

Quality Level

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 182.20

form

liquid

composition

Eugenol, 32.4%
Limonene, <0.1%
Linalool, 41.9%
Methyl chavicol, 0.9%

refractive index

n20/D 1.497

density

0.957 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

179.6 °F

flash_point_c

82 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Wan et al.
Journal of applied microbiology, 84(2), 152-158 (1998-06-20)
Basil essential oils, including basil sweet linalool (BSL) and basil methyl chavicol (BMC), were screened for antimicrobial activity against a range of Gram-positive and Gram-negative bacteria, yeasts and moulds using an agar well diffusion method. Both essential oils showed antimicrobial
K J Lachowicz et al.
Letters in applied microbiology, 26(3), 209-214 (1998-05-07)
Essential oils extracted by hydrodistillation from five different varieties of Ocimum basilicum L. plants (Anise, Bush, Cinnamon, Dark Opal and a commercial sample of dried basil) were examined for antimicrobial activity against a wide range of foodborne Gram-positive and -negative
Guy Kisluk et al.
Environmental microbiology, 15(10), 2787-2798 (2013-05-08)
The number of outbreaks of food-borne illness associated with consumption of fresh products has increased. A recent and noteworthy outbreak occurred in 2007. Basil contaminated with Salmonella enterica serovar Senftenberg was the source of this outbreak. Since basil produces high
Rahul Jain et al.
Drug development and industrial pharmacy, 34(4), 384-389 (2008-04-11)
The present work investigates effectiveness of basil oil, a volatile oil containing alcoholic terpenes, as a potential penetration enhancer for improved skin permeation of labetolol hydrochloride (LHCl) with reference to camphor, geraniol, thymol, and clove oil. Saturation solubilities of LHCl
V M Androsova
Tsitologiia, 28(9), 1030-1031 (1986-09-01)
Eugenol basil oil and eugenol are proposed as solvents of dye-stuffs in combined histological staining. These two substances were found to be of full value and more available substitutes of the oil of cloves.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service