Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W211710

Sigma-Aldrich

Peru balsam oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2117
EC Number:
Council of Europe no.:
298n
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Myroxylon pereirae Klotzsch

grade

Kosher

purified by

distillation

refractive index

n20/D 1.5690 (lit.)

bp

314 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

cinnamon; balsamic; sweet; vanilla

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: solvent extraction

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Nicolai et al.
Science immunology, 5(43) (2020-01-05)
During industrialization, humans have been exposed to increasing numbers of foreign chemicals. Failure of the immune system to tolerate drugs, cosmetics, and other skin products causes allergic contact dermatitis, a T cell-mediated disease with rising prevalence. Models of αβ T
Stephanie K Fabbro et al.
Current allergy and asthma reports, 14(10), 463-463 (2014-08-26)
Systemic contact dermatitis (SCD), a cutaneous reaction that is a direct manifestation of systemic exposure to a known allergen in a sensitized individual, has been increasingly recognized as a cause of persistent cutaneous contact dermatitis that is refractory to conventional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service