Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W209417

Sigma-Aldrich

Anise seed oil

Certified organic (NOP)

Synonym(s):

Anise oil, Anise seed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2094
Beilstein/REAXYS Number:
629884
MDL number:
UNSPSC Code:
12164502

biological source

Pimpinella anisum

grade

Certified organic (NOP)
Kosher

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 182.20

refractive index

n20/D 1.554 (lit.)

bp

232 °C (lit.)

density

0.980 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

anise

Looking for similar products? Visit Product Comparison Guide

General description

Anise seed oil is an essential oil extracted from the plant, Pimpinella anisum. It is mainly used in as flavoring and fragrance ingredient and also in traditional medicine due to its therapeutic properties. It generally contains trans-anethole and γ-himachalene as the key components.

Legal Information

Certified organic by MOSA

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essential oil content and composition of aniseed.
Acimovic MG, et al.
Zbornik Matice srpske za prirodne nauke, 128, 67-75 (2015)
Review of pharmacological properties and chemical constituents of Pimpinella anisum.
Shojaii A & Abdollahi Fard M.
ISRN pharmaceutics (2012)
Screening of antioxidant and antimicrobial activities of anise (Pimpinella anisum L.) seed extracts.
Gulc?n ?, et al.
Food Chemistry, 83(3), 371-382 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service