Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W207519

Sigma-Aldrich

Isoamyl hexanoate, mixture of isomers

natural, ≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22O2
CAS Number:
Molecular Weight:
186.29
FEMA Number:
2075
Council of Europe no.:
320
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.070

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

assay

≥98%

refractive index

n20/D 1.42 (lit.)

bp

222 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

apple; green; pineapple; sweet

SMILES string

CCCCCC(=O)OCCC(C)C

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service