W206504
α-Amylcinnamyl alcohol
mixture with Amyl hydrocinnamyl alcohol
Synonym(s):
2-Benzylidene-1-heptanol
About This Item
Recommended Products
biological source
synthetic
agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
FDA 21 CFR 172.515
composition
α-Amylcinnamyl alcohol, 25-50%
Amyl hydrocinnamyl alcohol, 50-75%
contains EU 1223/2009 and IFRA restricted α-Amyl Cinnamic Alcohol and α-Amyl Cinnamic Aldehyde
refractive index
n20/D 1.519 (lit.)
bp
141-143 °C/5 mmHg (lit.)
density
0.952 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
organoleptic
alcohol; jasmine
SMILES string
CCCCC\C(CO)=C/c1ccccc1
InChI
1S/C14H20O/c1-2-3-5-10-14(12-15)11-13-8-6-4-7-9-13/h4,6-9,11,15H,2-3,5,10,12H2,1H3/b14-11+
InChI key
LIPHCKNQPJXUQF-SDNWHVSQSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
>230.0 °F
flash_point_c
> 110 °C
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service