Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W202908

Sigma-Aldrich

Allyl 2-ethylbutyrate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2CHCO2CH2CH=CH2
CAS Number:
Molecular Weight:
156.22
FEMA Number:
2029
EC Number:
Council of Europe no.:
281
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.410

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

assay

≥98%

refractive index

n20/D 1.422 (lit.)

bp

165-167 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

oily; fruity; ethereal

SMILES string

CCC(CC)C(=O)OCC=C

InChI

1S/C9H16O2/c1-4-7-11-9(10)8(5-2)6-3/h4,8H,1,5-7H2,2-3H3

InChI key

NBKXNUWCFMZFMM-UHFFFAOYSA-N

Packaging

Packaged in glass bottles

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

129.2 °F - closed cup

flash_point_c

54 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service