Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W200816

Sigma-Aldrich

Acetoin

≥96%, natural

Synonym(s):

3-Hydroxy-2-butanone, Acetylmethylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(OH)CH3
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2008
Beilstein/REAXYS Number:
385636
EC Number:
Council of Europe no.:
749c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.051

grade

Kosher
natural

assay

≥96%

refractive index

n20/D 1.417 (lit.)

bp

148 °C (lit.)

mp

15 °C (monomer)
90 °C (dimer) (lit.)

density

1.013 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

creamy; dairy; fatty; milk; buttery

SMILES string

CC(O)C(C)=O

InChI

1S/C4H8O2/c1-3(5)4(2)6/h3,5H,1-2H3

InChI key

ROWKJAVDOGWPAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

replaced by

Product No.
Description
Pricing

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

118.4 °F - closed cup

flash_point_c

48 °C - closed cup

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jhonatan A Hernandez-Valdes et al.
Frontiers in microbiology, 11, 1032-1032 (2020-06-12)
Some secondary metabolites of fermentative bacteria are desired compounds for the food industry. Examples of these compounds are diacetyl and acetaldehyde, which are produced by species of the lactic acid bacteria (LAB) family. Diacetyl is an aromatic compound, giving the
Zhen Liu et al.
Bioresource technology, 102(22), 10741-10744 (2011-09-29)
Production of highly pure (2S,3S)-2,3-butanediol ((2S,3S)-2,3-BD) and (3S)-acetoin ((3S)-AC) in high concentrations is desirable but difficult to achieve. In the present study, glucose was first transformed to a mixture of (2S,3S)-2,3-BD and meso-2,3-BD by resting cells of Klebsiella pneumoniae CICC
Pieter Moons et al.
Applied and environmental microbiology, 77(10), 3422-3427 (2011-03-29)
During fermentation of sugars, a number of bacterial species are able to switch from mixed acid production to acetoin and 2,3-butanediol production in order to avoid lethal acidification of their environment, although the regulation of this switch is only poorly
M Otsuka et al.
Journal of chromatography, 577(2), 215-220 (1992-06-10)
Acetoin was quantitatively oxidized into diacetyl by Fe3+ in 1 M perchloric acid. The reaction of diacetyl with 4,5-dichloro-1,2-diaminobenzene afforded 6,7-dichloro-2,3-dimethylquinoxaline (DCDMQ), which was extracted by benzene containing aldrin (25 ng/ml) as an internal standard, and determined by gas chromatography
Ranjita Biswas et al.
Applied microbiology and biotechnology, 94(3), 651-658 (2012-03-01)
Production of 2,3-butanediol by Bacillus subtilis takes place in late-log or stationary phase, depending on the expression of bdhA gene encoding acetoin reductase, which converts acetoin to 2,3-butanediol. The present work focuses on the development of a strain of B.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service