Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W200794

Sigma-Aldrich

Triacetin

≥99.5%

Synonym(s):

1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glyceryl triacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COOCH2)2CHOCOCH3
CAS Number:
Molecular Weight:
218.20
FEMA Number:
2007
Beilstein/REAXYS Number:
1792353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

7.52 (vs air)

assay

≥99.5%

autoignition temp.

809 °F

expl. lim.

1.05 %, 189 °F
7.73 %, 215 °F

refractive index

n25/D 1.429-1.431 (lit.)

bp

258 °C (lit.)

mp

3 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCC(COC(C)=O)OC(C)=O

InChI

1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3

InChI key

URAYPUMNDPQOKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Legal Information

Manufactured by Eastman Chemical Company. Distributed by SAFC.

comparable product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

298.4 °F - closed cup

flash_point_c

148 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hui Liu et al.
Journal of pharmaceutical sciences, 101(5), 1783-1793 (2012-02-10)
Although injectable depot-forming solutions have been commercialized, the factors that influence the overall release kinetics from such systems are still not fully understood. In this work, we address the effect of cosolvent on the issue of excessive burst release of
Seung Jun Choi et al.
Journal of agricultural and food chemistry, 57(23), 11349-11353 (2009-11-07)
Citral is widely used in the beverage, food, and fragrance industries for its characteristic flavor profile. However, it chemically degrades over time in aqueous solutions due to an acid-catalyzed reaction, which leads to loss of desirable flavor notes and formation
Huayin Pu et al.
Journal of agricultural and food chemistry, 59(10), 5738-5745 (2011-04-26)
An oral colon-targeting controlled release system based on resistant starch acetate (RSA) as a film-coating material was developed. The RSA was successfully synthesized, and its digestion resistibility could be improved by increasing the degree of substitution (DS), which was favorable
Nilufer Yuksel et al.
International journal of pharmaceutics, 404(1-2), 102-109 (2010-11-26)
The purpose of this study was to form indomethacin (IND)-loaded poly(methyl methacrylate) (PMMA) microspheres having an extended drug release profile over a period of 24h. Microspheres were prepared by solvent evaporation method using sucrose stearate as a droplet stabilizer. When
C N Madhavarao et al.
Journal of inherited metabolic disease, 32(5), 640-640 (2009-08-18)
Canavan disease (CD) is a fatal dysmyelinating genetic disorder associated with aspartoacylase deficiency, resulting in decreased brain acetate levels and reduced myelin lipid synthesis in the developing brain. Here we tested tolerability of a potent acetate precursor, glyceryl triacetate (GTA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service