Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W200409

Sigma-Aldrich

Acetaldehyde phenethyl propyl acetal

≥95%

Synonym(s):

Leafy acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OCH2CH2C6H5)OCH2CH2CH3
CAS Number:
Molecular Weight:
208.30
FEMA Number:
2004
EC Number:
Council of Europe no.:
511
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.016
NACRES:
NA.21
organoleptic:
green; leafy; ethereal
biological source:
synthetic

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

assay

≥95%

may contain

<0.20% BHT, synthetic as stabilizer

refractive index

n20/D 1.479 (lit.)

density

0.951 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

green; leafy; ethereal

SMILES string

CCCOC(C)OCCc1ccccc1

InChI

1S/C13H20O2/c1-3-10-14-12(2)15-11-9-13-7-5-4-6-8-13/h4-8,12H,3,9-11H2,1-2H3

InChI key

FMYCPRQGKSONCP-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

206.6 °F - closed cup

flash_point_c

97.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service