Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V319902

Sigma-Aldrich

Ethyl acetate

ACS reagent

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein/REAXYS Number:
506104
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
ACS reagent
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

grade

ACS reagent

Quality Level

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

dilution

(for analytical testing)

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Amanda Holm Rottbøll et al.
Basic & clinical pharmacology & toxicology, 117(4), 242-250 (2015-04-02)
Recent intrabronchial microdialysis data indicate that the respiratory epithelium is highly permeable to drugs. Of concern is whether intrabronchial microdialysis disrupts the integrity of the respiratory epithelium and thereby alters drug penetration into the pulmonary epithelial lining fluid (PELF). The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service