Key Documents
THSX1141
ThalesNano X-Cube CatCart® catalyst cartridge system
azide, polymer-bound
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
UNSPSC Code:
41103817
NACRES:
NB.51
manufacturer/tradename
Nanotechnologiai Rt.
General description
CatCarts are suitable for use only with Thalesnano X-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
X-Cube is a trademark of ThalesNano Nanotechnology Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)