Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

THSX1134

ThalesNano X-Cube CatCart® catalyst cartridge system

AlCl3-functionalized silica

Synonym(s):

Heterogeneous catalysis system, safe Heterogeneous catalysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103817

manufacturer/tradename

Nanotechnologiai Rt.

Looking for similar products? Visit Product Comparison Guide

General description

CatCarts are suitable for use only with Thalesnano X-Cube reactors.

Legal Information

CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
X-Cube is a trademark of ThalesNano Nanotechnology Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service