THS05142
ThalesNano MicroCatCart™ catalyst cartridge system
L × diam. 30 mm × 4 mm, Pd EnCat® TPP 30
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103817
packaging
pack of 1 ea
manufacturer/tradename
ThalesNano THS 05142
L × diam.
30 mm × 4 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
EnCat is a registered trademark of Reaxa Ltd.
MicroCatCart is a trademark of ThalesNano Nanotechnology Inc.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service