Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

THS03141

ThalesNano MicroCatCart catalyst cartridge system

L × diam. 30 mm × 4 mm, NP Pd(0) EnCat® 30

Synonym(s):

Heterogeneous catalysis system, safe Heterogeneous catalysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103817
NACRES:
NB.51

packaging

pkg of 6 ea

manufacturer/tradename

ThalesNano THS 03141

L × diam.

30 mm × 4 mm

Looking for similar products? Visit Product Comparison Guide

Legal Information

EnCat is a registered trademark of Reaxa Ltd.
MicroCatCart is a trademark of ThalesNano Nanotechnology Inc.

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation

target_organs

Nervous system

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service