Key Documents
THS01811
ThalesNano CatCart® catalyst cartridge system, 30 mm L
Pd, sulfided
About This Item
UNSPSC Code:
41115709
diam.
4 mm
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)