Key Documents
TEM-LC400NIUL
Sigma-Aldrich
Ultrathin Lacey Carbon Supported Nickel Grid
size 400 mesh, box of 25
Synonym(s):
LACEY CARBON FILM - NI 400 MESH ULTRA THIN 25/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 25
size
400 mesh
General description
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Nickel Grid, 400 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)