Key Documents
TEM-HC300CU100
Sigma-Aldrich
Holey Carbon Supported Copper Grids
size 300 mesh, size 100 nm , Hole, box of 25
Synonym(s):
Holey Carbon - Cu, 300 mesh, 100 micron
About This Item
UNSPSC Code:
41111761
material
carbon film (holey)
copper grid
packaging
box of 25
size
100 nm , Hole
300 mesh
General description
Holey carbon film supported copper TEM gird: hole size 100 nm on 300 mesh copper grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)