Key Documents
TEM-G200MO
Sigma-Aldrich
Glider Grids Standard Molybdenum Square Mesh
size 200 mesh, vial of 25
Synonym(s):
MOLYBDENUM GRIDS 200#
About This Item
UNSPSC Code:
41111761
packaging
vial of 25
size
200 mesh
General description
Gilder Grids Standard Square Mesh: Molybdenum Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 125μm, Hole 90μm, Bar 35μm
Choose from one of the most recent versions:
Certificates of Analysis (COA)