Key Documents
TEM-G200AU
Sigma-Aldrich
Glider Grids Standard Gold Square Mesh
size 200 mesh, vial of 50
Synonym(s):
SQUARE 200 MESH GOLD 50/VIAL
About This Item
UNSPSC Code:
41111761
packaging
vial of 50
size
200 mesh
General description
Gilder Grids Standard Square Mesh: Gold Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 125μm, Hole 90μm, Bar 35μm
Choose from one of the most recent versions:
Certificates of Analysis (COA)