Key Documents
TEM-FFT300CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Thin Bar Square Mesh
size 300 mesh, box of 50
Synonym(s):
FORMVAR THIN BAR SQUARE MESH - CU 300 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
300 mesh
General description
A 5-6 nm thin film of pure formvar resin supported thin bar square mesh grids. Copper grid, 300 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)