Key Documents
TEM-FF200CU
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 200 mesh, box of 25
Synonym(s):
FORMVAR 200 MESH CU 25/BX
About This Item
UNSPSC Code:
41111761
material
copper grid
formvar film
description
film layer thickness 10 nm
packaging
box of 25
size
200 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 200 mesh. All of our film is laid on the shiny side of the grid.
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)