Key Documents
TEM-FF100CU
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 100 mesh, box of 25
Synonym(s):
FORMVAR FILM 100# COPPER 25/VIAL
About This Item
UNSPSC Code:
41111761
packaging
box of 25
size
100 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 100 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)