Key Documents
TEM-FF100CU50
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 100 mesh, box of 50
Synonym(s):
FORMVAR FILM 100 MESH CU 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
100 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 100 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)