Key Documents
TEM-FCF150NI
Sigma-Aldrich
Formvar/Carbon Supported Nickel Square Mesh
size 150 mesh, box of 25
Synonym(s):
FORMVAR/CARBON 150 NI 25/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 25
size
150 mesh
General description
A 10nm formvar and 1nm carbon film supported square mesh grid. Nickel grid, 150 mesh. A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)