Key Documents
TEM-FCF150CUUA
Sigma-Aldrich
Ultrathin Formvar/Carbon Supported Copper Square Mesh
size 150 mesh, box of 50
Synonym(s):
FORMVAR/CARBON SQUARE MESH -150 MESH UA 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
150 mesh
General description
Ultrathin 5-6nm Formvar and 1nm Carbon Supported Square Mesh Grids: Copper Grid, 150 mesh. A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)